Learn More
Aviva Systems Biology Corp TGM2 Antibody - middle region (ARP47471_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47471_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TGM2. Alternate names: TG2, TGC, GNAH, G-ALPHA-hApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59560