Learn More
Aviva Systems Biology Corp TGM5 Antibody - C-terminal region (ARP49208_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49208_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGM5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TGM5. Alternate names: MGC141907, TGM6, TGMX, TGX, TGASE5, TGASEXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60042