Learn More
Aviva Systems Biology Corp TGM6 Antibody - middle region (ARP49194_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49194_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGM6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TGM6. Alternate names: TGY, TGM3L, dJ734P14.3, TG6, SCA35ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60038