Learn More
Medchemexpress LLC TGN-020 sodium | 1313731-99-5 | 100.0% | 228.21 | 250 MG
SDP
Catalog No. 5000451286
Encompass_Preferred
TGN-020 sodium is a selective Aquaporin 4 (AQP4) inhibitor with an IC50 of 3.1 μM. It is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. This compound has been shown to alleviate edema and inhibit glial scar formation following spinal cord compression injury in rats.
  • Selective Aquaporin 4 (AQP4) inhibitor.
  • Acts as an alkyl chain-based PROTAC linker for PROTAC synthesis.
  • Alleviates edema in rat models of spinal cord compression injury.
  • Inhibits glial scar formation in rat models of spinal cord compression injury.
  • Suppresses retinal edema in STZ-induced diabetic rats.
Catalog No. 50-004-51286 Supplier Medchemexpress LLC Supplier No. 250MGHYW008574A
TGN-020 sodium is a selective Aquaporin 4 (AQP4) inhibitor with an IC50 of 3.1 μM. It is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. This compound has been shown to alleviate edema and inhibit glial scar formation following spinal cord compression injury in rats.
  • Selective Aquaporin 4 (AQP4) inhibitor.
  • Acts as an alkyl chain-based PROTAC linker for PROTAC synthesis.
  • Alleviates edema in rat models of spinal cord compression injury.
  • Inhibits glial scar formation in rat models of spinal cord compression injury.
  • Suppresses retinal edema in STZ-induced diabetic rats.