Learn More
Cayman Chemical Tigogenin, Purity: ≥90%, CAS: 77-60-1, 100 mg
SDP
Catalog No. 502990553
Encompass_Preferred
A steroidal sapogenin with diverse biological activities; increases the proliferation rate of, as well as mRNA levels of genes encoding the osteoblastic differentiation markers Cbfa1, collagen type I, and osteocalcin in, mouse BMSCs in a concentration-dependent manner; increases matrix calcium deposition in BMSCs at 30 and 90 μM; active against the fungus A. fumigatus (MIC50 = 16 μg/ml); inhibits carrageenan-induced paw edema in rats at 4.2 μg/kg
Catalog No. 50-299-0553 Supplier Cayman Chemical Supplier No. 30137100 MG
A steroidal sapogenin with diverse biological activities; increases the proliferation rate of, as well as mRNA levels of genes encoding the osteoblastic differentiation markers Cbfa1, collagen type I, and osteocalcin in, mouse BMSCs in a concentration-dependent manner; increases matrix calcium deposition in BMSCs at 30 and 90 μM; active against the fungus A. fumigatus (MIC50 = 16 μg/ml); inhibits carrageenan-induced paw edema in rats at 4.2 μg/kg