Learn More
Aviva Systems Biology Corp TGOLN2 Antibody - N-terminal region (ARP46727_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46727_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGOLN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TGOLN2. Alternate names: MGC14722, TGN38, TGN46, TGN48, TGN51, TTGN2ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59384