Learn More
Eurofins Discoverx TGR5 Receptor Agonist, 5 mg. (92-1196)
Catalog No. 501999724
Encompass
TGR5 Receptor Agonist is a synthetic cell-permeable small molecule agonist for the G protein-coupled bile acid receptor TGR5 (pEC50=6.8-7.5 nM). Improves glucagon like peptide-1 (GLP-1) secretion from intestinal cells, and mediates energy balance, inflammation, and digestion. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9724 Supplier Eurofins Discoverx Supplier No. 921196
TGR5 Receptor Agonist is a synthetic cell-permeable small molecule agonist for the G protein-coupled bile acid receptor TGR5 (pEC50=6.8-7.5 nM). Improves glucagon like peptide-1 (GLP-1) secretion from intestinal cells, and mediates energy balance, inflammation, and digestion. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.