Learn More
Selleck Chemical LLC TGR5 Receptor Agonist S0193-25MG
SDP
Catalog No. 5000040203
Encompass_Preferred
TGR5 Receptor Agonist (TGR5 CCDC MDK00245 compd 6) is a potent agonist of TGR5(GPCR19 GPBAR1 M-BAR) TGR5 Receptor Agonist shows improved potency with pEC50 of 6 8 and 7 5 in the U2-OS cell assay and in melanophore cells respectively
Catalog No. 50-000-40203 Supplier Selleck Chemical LLC Supplier No. S019325MG
TGR5 Receptor Agonist (TGR5 CCDC MDK00245 compd 6) is a potent agonist of TGR5(GPCR19 GPBAR1 M-BAR) TGR5 Receptor Agonist shows improved potency with pEC50 of 6 8 and 7 5 in the U2-OS cell assay and in melanophore cells respectively