Learn More
Medchemexpress LLC HY-14229 5mg Medchemexpress, TGR5 Receptor Agonist CAS:1197300-24-5 Purity:>98%
SDP
Catalog No. 502081575
Encompass_Preferred
Medchemexpress, HY-14229 5mg TGR5 Receptor Agonist CAS:1197300-24-5 TGR5 Receptor Agonist (CCDC), a potent TGR5(GPCR19) agonist, shows improved potency in the U2-OS cell assay (pEC50=6.8) and in melanophore cells (pEC50=7.5). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1575 Supplier Medchemexpress LLC Supplier No. HY142295MG
Medchemexpress, HY-14229 5mg TGR5 Receptor Agonist CAS:1197300-24-5 TGR5 Receptor Agonist (CCDC), a potent TGR5(GPCR19) agonist, shows improved potency in the U2-OS cell assay (pEC50=6.8) and in melanophore cells (pEC50=7.5). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.