Learn More
Cayman Chemical TGR5GP-BAR1 Reporter asy 1ea
SDP
Catalog No. 502892605
Encompass_Preferred
An IRE1a inhibitor (IC50s 0.2 and 0.02 UM for RNase and kinase activity respectively) selective for IRE1a over Ron and FGFR1 (IC50s 4.4 and 17 UM) as well as a panel of 282 kinases at 10 UM inhibits ER stress-induced increases in the splicing of mRNA encoding XBP1 in eight multiple myeloma cell lines at 8 UM reduces viral replication in tick-borne encephalitis virus-infected SH-SY5Y and SNB-19 cells at 0.5 UM and tick-borne encephalitis virus-infected Caco-2 cells at 1 UM
Catalog No. 50-289-2605 Supplier Cayman Chemical Supplier No. 6014401 EA
An IRE1a inhibitor (IC50s 0.2 and 0.02 UM for RNase and kinase activity respectively) selective for IRE1a over Ron and FGFR1 (IC50s 4.4 and 17 UM) as well as a panel of 282 kinases at 10 UM inhibits ER stress-induced increases in the splicing of mRNA encoding XBP1 in eight multiple myeloma cell lines at 8 UM reduces viral replication in tick-borne encephalitis virus-infected SH-SY5Y and SNB-19 cells at 0.5 UM and tick-borne encephalitis virus-infected Caco-2 cells at 1 UM