Learn More
AG Scientific Inc TGS Buffer 10X Solution 1 L
Catalog No. 502268409
Encompass_Preferred
TRIS-Glycine-SDS is a common buffer used to run SDS-PAGE gels in electrophoresis. TGS helps maintain the pH and temperature of the environment and provides ions to carry electrical current. It is usually used as the anode and cathode buffer.Recommended to run at 150 V for mini vertical gel electrophoresis units.
Catalog No. 50-226-8409 Supplier AG Scientific Inc Supplier No. T27111L
TRIS-Glycine-SDS is a common buffer used to run SDS-PAGE gels in electrophoresis. TGS helps maintain the pH and temperature of the environment and provides ions to carry electrical current. It is usually used as the anode and cathode buffer.Recommended to run at 150 V for mini vertical gel electrophoresis units.