Learn More
Aviva Systems Biology Corp TGS1 Antibody - middle region (ARP48923_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48923_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TGS1. Alternate names: DKFZp762A163, FLJ22995, NCOA6IP, PIMT, PIPMTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59962