Learn More
Bioworld Tris-Glycine (TG) Running Buffer (10X), 4 L
Catalog No. 50196711
Encompass_Preferred
Product DescriptionTris-Glycine (TG) running buffer has a concentration of 10X with a pH of 8.3. 1X solution contains 0.025M Tris base and 0.192M Glycine. Product is of ultra pure grade.TG buffers are blended using pure reagents to provide high quality for protein research. This buffer is formulated using bioPlus Tris powder to ensure exceptional purity along with the absence of nuclease and protease activities. TG Buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes.
Catalog No. 50-196-711 Supplier Bioworld Supplier No. 105300312
Product DescriptionTris-Glycine (TG) running buffer has a concentration of 10X with a pH of 8.3. 1X solution contains 0.025M Tris base and 0.192M Glycine. Product is of ultra pure grade.TG buffers are blended using pure reagents to provide high quality for protein research. This buffer is formulated using bioPlus Tris powder to ensure exceptional purity along with the absence of nuclease and protease activities. TG Buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes.