Learn More
Bioworld Tris-Glycine (TG) Running Buffer, Ready-Pack (1X), 1 PK(s)
Catalog No. 50196712
Encompass_Preferred
Tris-Glycine The TG Buffers are blended using pure reagents to provide the highest quality buffers for protein research. Ideal for protein separation, this buffer is formulated using bioPlus Tris powder to ensure exceptional purity and the absence of nuclease and protease activities. TG Buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes. Technical Information:Uniform white powderUltra Pure GradeContents (1X solution): 0.025M Tris base and 0.192M Glycine.pH @25C: 8.3Each Ready-pack prepares 1L of 10X Concentrate.Supplied as 1x20 PacksStorage: RTHazard: Irritant
Catalog No. 50-196-712 Supplier Bioworld Supplier No. 105300331
Tris-Glycine The TG Buffers are blended using pure reagents to provide the highest quality buffers for protein research. Ideal for protein separation, this buffer is formulated using bioPlus Tris powder to ensure exceptional purity and the absence of nuclease and protease activities. TG Buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes. Technical Information:Uniform white powderUltra Pure GradeContents (1X solution): 0.025M Tris base and 0.192M Glycine.pH @25C: 8.3Each Ready-pack prepares 1L of 10X Concentrate.Supplied as 1x20 PacksStorage: RTHazard: Irritant