Learn More
Medchemexpress LLC HY-10114 10mg , TGX-221 CAS:663619-89-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011410MG
Encompass_Preferred
HY-10114 10mg TGX-221 CAS: 663619-89-4 TGX-221 is a potent, selective, and cell membrane permeable inhibitor of the PI3K p110β catalytic subunit, used for cancer treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0392