Learn More
Enzo Life Sciences TGX-221 (1mg). CAS: 663619-89-4
SDP
Supplier:  Enzo Life Sciences ALX270464M001
Encompass_Preferred
Potent, selective and cell permeable inhibitor of phosphoinositide 3-kinase (PI(3)K) p110β. Alternative name: 7-Methyl-2-(4-morpholinyl)-9-[1-(phenylamino)ethyl]-4H-pyrido-[1,2-a]pyrimidine-4-one. Purity: ≥98% (NMR). Solubility: Soluble in DMSO, 100% ethanol or dimethyl formamide; sparingly soluble in aqueous buffers. Long Term Storage: -20°C.
Catalog No. 50-200-8586