Learn More
Medchemexpress LLC Th-263 | 313520-94-4 | 99.0% | 380.46 g·mol⁻¹ | C21H20N2O3S | 5 MG
SDP
Catalog No. 5000223109
Encompass_Preferred
TH-263 is a diaryl sulfonamide derivative used as a negative control for LIM kinase inhibitor studies. It is reported to be inactive against LIMK1 and LIMK2 and is supplied as a high-purity solid for in vitro laboratory research.
  • Diaryl sulfonamide structure suitable as a negative control.
  • Inactive toward LIMK1 and LIMK2 in reported assays.
  • 99.0% purity for consistent experimental results.
  • White to off-white solid appearance.
  • Stable when stored as powder at -20°C; in solvent long-term storage recommended at -80°C.
Catalog No. 50-002-23109 Supplier Medchemexpress LLC Supplier No. HY1358435MG
TH-263 is a diaryl sulfonamide derivative used as a negative control for LIM kinase inhibitor studies. It is reported to be inactive against LIMK1 and LIMK2 and is supplied as a high-purity solid for in vitro laboratory research.
  • Diaryl sulfonamide structure suitable as a negative control.
  • Inactive toward LIMK1 and LIMK2 in reported assays.
  • 99.0% purity for consistent experimental results.
  • White to off-white solid appearance.
  • Stable when stored as powder at -20°C; in solvent long-term storage recommended at -80°C.