Learn More
Medchemexpress LLC HY-10535 100mg , TH-302 Evofosfamide CAS:918633-87-1 Purity:98%
SDP
Catalog No. 501492960
Encompass_Preferred
Medchemexpress, HY-10535 100mg TH-302 Evofosfamide CAS:918633-87-1 Formula:C9H16Br2N5O4P Hypoxia-activated prodrug Purity:98% TH-302 is a hypoxia -activated prodrug with IC 50 of 10 μM and 1000 μM in hypoxia (N 2 ) and normoxia (21% O 2 ), respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2960 Supplier Medchemexpress LLC Supplier No. HY10535100MG
Medchemexpress, HY-10535 100mg TH-302 Evofosfamide CAS:918633-87-1 Formula:C9H16Br2N5O4P Hypoxia-activated prodrug Purity:98% TH-302 is a hypoxia -activated prodrug with IC 50 of 10 μM and 1000 μM in hypoxia (N 2 ) and normoxia (21% O 2 ), respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.