Learn More
Medchemexpress LLC Th-6 | 3031349-25-1 | 95.0% | 429.44 | 1 ML
SDP
Catalog No. 5000360931
Encompass_Preferred
TH-6 is a potent HDAC inhibitor with IC50s of 0.115, 0.135, 0.242, 0.138, and 2.120 μM for HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8, respectively. It demonstrates anti-tumor activity and inhibits cell migration and invasion, inducing apoptosis and cell cycle arrest at the G2/M phase.
  • Inhibits cell migration and invasion
  • Induces apoptosis
  • Causes cell cycle arrest at the G2/M phase
  • Demonstrates anti-tumor activity
  • Potent HDAC inhibitor
Catalog No. 50-003-60931 Supplier Medchemexpress LLC Supplier No. HY14902910MM/1ML
TH-6 is a potent HDAC inhibitor with IC50s of 0.115, 0.135, 0.242, 0.138, and 2.120 μM for HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8, respectively. It demonstrates anti-tumor activity and inhibits cell migration and invasion, inducing apoptosis and cell cycle arrest at the G2/M phase.
  • Inhibits cell migration and invasion
  • Induces apoptosis
  • Causes cell cycle arrest at the G2/M phase
  • Demonstrates anti-tumor activity
  • Potent HDAC inhibitor