Learn More
Medchemexpress LLC TH-6 | 3031349-25-1 | 95.0% | 429.44 | 5 MG
SDP
Catalog No. 5000360932
Encompass_Preferred
TH-6 is a potent HDAC inhibitor that targets HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8. This compound has demonstrated anti-tumor activity by inhibiting cell migration and invasion, and inducing apoptosis and cell cycle arrest at the G2/M phase.
  • Potent HDAC inhibitor
  • Inhibits cell migration and invasion
  • Induces apoptosis
  • Induces cell cycle arrest at G2/M phase
  • Demonstrates anti-tumor activity
  • For research use only
Catalog No. 50-003-60932 Supplier Medchemexpress LLC Supplier No. HY1490295MG
TH-6 is a potent HDAC inhibitor that targets HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8. This compound has demonstrated anti-tumor activity by inhibiting cell migration and invasion, and inducing apoptosis and cell cycle arrest at the G2/M phase.
  • Potent HDAC inhibitor
  • Inhibits cell migration and invasion
  • Induces apoptosis
  • Induces cell cycle arrest at G2/M phase
  • Demonstrates anti-tumor activity
  • For research use only