Learn More
Medchemexpress LLC TH-6 | 3031349-25-1 | 95.0% | 429.44 | 50 MG
SDP
Catalog No. 5000359526
Encompass_Preferred
TH-6 is a potent HDAC inhibitor that shows anti-tumor activity. It inhibits cell migration and invasion, and induces apoptosis and cell cycle arrest at G2/M phase.
  • Potent HDAC inhibitor with specific IC50s for HDAC1, HDAC2, HDAC3, HDAC6, HDAC8.
  • Inhibits cell migration and invasion.
  • Induces apoptosis and cell cycle arrest at G2/M phase.
  • Shows anti-tumor activity in mice models.
  • Exhibits antiproliferative activities in various cancer cell lines and normal human lung cells.
  • Inhibits tubulin polymerization.
  • Increases expression of Ac-α-Tubulin and AC-Histone H3 in HepG2 cells.
  • Shows favorable liver microsomal stability in vitro.
Catalog No. 50-003-59526 Supplier Medchemexpress LLC Supplier No. HY14902950MG
TH-6 is a potent HDAC inhibitor that shows anti-tumor activity. It inhibits cell migration and invasion, and induces apoptosis and cell cycle arrest at G2/M phase.
  • Potent HDAC inhibitor with specific IC50s for HDAC1, HDAC2, HDAC3, HDAC6, HDAC8.
  • Inhibits cell migration and invasion.
  • Induces apoptosis and cell cycle arrest at G2/M phase.
  • Shows anti-tumor activity in mice models.
  • Exhibits antiproliferative activities in various cancer cell lines and normal human lung cells.
  • Inhibits tubulin polymerization.
  • Increases expression of Ac-α-Tubulin and AC-Histone H3 in HepG2 cells.
  • Shows favorable liver microsomal stability in vitro.