Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TH-6 | 3031349-25-1 | 95.0% | 429.44 | C22H24FN3O5 | 10 MG
SDP

Catalog No. 5000285568
Encompass_Preferred

TH-6 is a potent histone deacetylase (HDAC) inhibitor intended for research applications. It inhibits multiple HDAC isoforms, induces apoptosis, causes G2/M cell cycle arrest, and demonstrates antiproliferative and antitumor activity in preclinical models.

  • Potent HDAC inhibition with submicromolar to low micromolar IC50s.
  • Inhibits cell migration and invasion in cancer cell models.
  • Induces apoptosis and G2/M phase cell cycle arrest.
  • Inhibits tubulin polymerization (IC50 4.06 μM) and increases acetylated α-tubulin and histone H3 levels.
  • Supplied as a light yellow to khaki solid with 95.0% purity.
  • Molecular weight 429.44; formula C22H24FN3O5; CAS 3031349-25-1.
  • Soluble in DMSO at 50 mg/mL (requires warming and sonication).

Catalog No. 50-002-85568 Supplier Medchemexpress LLC Supplier No. HY14902910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TH-6 is a potent histone deacetylase (HDAC) inhibitor intended for research applications. It inhibits multiple HDAC isoforms, induces apoptosis, causes G2/M cell cycle arrest, and demonstrates antiproliferative and antitumor activity in preclinical models.

  • Potent HDAC inhibition with submicromolar to low micromolar IC50s.
  • Inhibits cell migration and invasion in cancer cell models.
  • Induces apoptosis and G2/M phase cell cycle arrest.
  • Inhibits tubulin polymerization (IC50 4.06 μM) and increases acetylated α-tubulin and histone H3 levels.
  • Supplied as a light yellow to khaki solid with 95.0% purity.
  • Molecular weight 429.44; formula C22H24FN3O5; CAS 3031349-25-1.
  • Soluble in DMSO at 50 mg/mL (requires warming and sonication).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.