Learn More
Aviva Systems Biology Corp TH Antibody - N-terminal region (ARP30889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human TH. Alternate names: TYH, DYT14, DYT5bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54170