Learn More
Aviva Systems Biology Corp TH Antibody - N-terminal region (AVARP01052_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP01052_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human TH. Alternate names: TYH, DYT14, DYT5bApplicationIHC, WB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62472