Learn More
Bellco Glass, Inc. TH Cap w/PTFE Silicone Liner 8-425mm
SDP
Catalog No. 501945382
Encompass_Preferred
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA.
Catalog No. 50-194-5382 Supplier Bellco Glass, Inc. Supplier No. 207000750
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA.