Learn More
Sino Biological Anti-Tyrosine Hydroxylase Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50997RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TH / Tyrosine Hydroxylase (rM TH / Tyrosine Hydroxylase; Catalog#50997-M07B; P24529; Pro 2-Ser 498). TH / Tyrosine Hydroxylase specific IgG was purified by Mouse TH / Tyrosine Hydroxylase affinity chromatography.
Catalog No. 50-228-2581