Learn More
Medchemexpress LLC Th-z145 | 2260887-57-6 | 97.1% | 394.34 g/mol | C16H28O7P2 | 10 MG
SDP
Catalog No. 5000286873
Encompass_Preferred
TH-Z145 is a lipophilic bisphosphonate research compound that inhibits farnesyl pyrophosphate synthase (FPPS) with a reported IC50 of 210 nM. The compound is characterized by defined physicochemical properties (CAS 2260887-57-6; formula C16H28O7P2; mol wt 394.34 g/mol) and is supplied for research use in small milligram quantities, typically as a DMSO-soluble solid.
  • Inhibits farnesyl pyrophosphate synthase with reported IC50 of 210 nM.
  • Lipophilic bisphosphonate scaffold suitable for biochemical assays.
  • High purity suitable for research applications.
  • Supplied in small milligram packages for screening and synthesis.
  • Soluble in DMSO for convenient stock preparation.
  • Includes datasheet and analytical information for verification.
Catalog No. 50-002-86873 Supplier Medchemexpress LLC Supplier No. HY15016810MG
TH-Z145 is a lipophilic bisphosphonate research compound that inhibits farnesyl pyrophosphate synthase (FPPS) with a reported IC50 of 210 nM. The compound is characterized by defined physicochemical properties (CAS 2260887-57-6; formula C16H28O7P2; mol wt 394.34 g/mol) and is supplied for research use in small milligram quantities, typically as a DMSO-soluble solid.
  • Inhibits farnesyl pyrophosphate synthase with reported IC50 of 210 nM.
  • Lipophilic bisphosphonate scaffold suitable for biochemical assays.
  • High purity suitable for research applications.
  • Supplied in small milligram packages for screening and synthesis.
  • Soluble in DMSO for convenient stock preparation.
  • Includes datasheet and analytical information for verification.