Learn More
Medchemexpress LLC TH-Z145 | 2260887-57-6 | C16H28O7P2 | 1 ML
SDP
Catalog No. 5000375085
Encompass_Preferred
TH-Z145 is a lipophilic bisphosphonate and an FPPS inhibitor with an IC50 of 210 nM. It has shown strong prophylactic effects in a pathogenic influenza model and inhibits tumor growth, prolonging survival in a B16-OVA cells xenograft mice model. It is intended for research use only.
  • Lipophilic bisphosphonate and FPPS inhibitor
  • IC50 of 210 nM
  • Strong prophylactic effects in pathogenic influenza model
  • Inhibits tumor growth and prolongs survival in B16-OVA cells xenograft mice model
Catalog No. 50-003-75085 Supplier Medchemexpress LLC Supplier No. HY15016810MM/1ML
TH-Z145 is a lipophilic bisphosphonate and an FPPS inhibitor with an IC50 of 210 nM. It has shown strong prophylactic effects in a pathogenic influenza model and inhibits tumor growth, prolonging survival in a B16-OVA cells xenograft mice model. It is intended for research use only.
  • Lipophilic bisphosphonate and FPPS inhibitor
  • IC50 of 210 nM
  • Strong prophylactic effects in pathogenic influenza model
  • Inhibits tumor growth and prolongs survival in B16-OVA cells xenograft mice model