Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Th-Z145 | 2260887-57-6 | C16H28O7P2 | 5 MG
SDP

Catalog No. 5000375240
Encompass_Preferred

TH-Z145 is a lipophilic bisphosphonate that acts as an FPPS inhibitor. It has demonstrated strong prophylactic effects in a pathogenic influenza model and inhibited tumor growth in a xenograft mice model. It appears as an off-white to light yellow solid with a purity of 99.69%.

  • FPPS inhibitor with an IC50 of 210 nM
  • Demonstrated strong prophylactic effects in pathogenic influenza model
  • Inhibited tumor growth and prolonged survival in xenograft mice model
  • Soluble in DMSO at 100 mg/mL
  • Purity of 99.69%

Catalog No. 50-003-75240 Supplier Medchemexpress LLC Supplier No. HY1501685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TH-Z145 is a lipophilic bisphosphonate that acts as an FPPS inhibitor. It has demonstrated strong prophylactic effects in a pathogenic influenza model and inhibited tumor growth in a xenograft mice model. It appears as an off-white to light yellow solid with a purity of 99.69%.

  • FPPS inhibitor with an IC50 of 210 nM
  • Demonstrated strong prophylactic effects in pathogenic influenza model
  • Inhibited tumor growth and prolonged survival in xenograft mice model
  • Soluble in DMSO at 100 mg/mL
  • Purity of 99.69%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.