Learn More
Medchemexpress LLC TH-Z145 | 2260887-57-6 | C16H28O7P2 | 50 MG
SDP
Catalog No. 5000375239
Encompass_Preferred
TH-Z145 is a lipophilic bisphosphonate that functions as an FPPS inhibitor with an IC50 of 210 nM. It is intended for research applications only.
  • Acts as an FPPS inhibitor
  • Exhibits strong prophylactic effects in pathogenic influenza models
  • Inhibits tumor growth and prolongs survival in xenograft mice models
Catalog No. 50-003-75239 Supplier Medchemexpress LLC Supplier No. HY15016850MG
TH-Z145 is a lipophilic bisphosphonate that functions as an FPPS inhibitor with an IC50 of 210 nM. It is intended for research applications only.
  • Acts as an FPPS inhibitor
  • Exhibits strong prophylactic effects in pathogenic influenza models
  • Inhibits tumor growth and prolongs survival in xenograft mice models