Learn More
Medchemexpress LLC TH-Z93 | 2260887-09-8 | C12H22N2O7P2 | 5 MG
SDP
Catalog No. 5000375006
Encompass_Preferred
TH-Z93 is a lipophilic bisphosphonate that acts as an FPPS inhibitor with an IC50 of 90 nM. It exhibits strong prophylactic effects in a pathogenic influenza model, inhibiting tumor growth and prolonging survival in a B16-OVA cells (s.c.) xenograft mice model. It is for research use only.
  • FPPS inhibitor with an IC50 of 90 nM
  • Exhibits strong prophylactic effects in pathogenic influenza model
  • Inhibits tumor growth and prolongs survival in xenograft mice model
Catalog No. 50-003-75006 Supplier Medchemexpress LLC Supplier No. HY1501675MG
TH-Z93 is a lipophilic bisphosphonate that acts as an FPPS inhibitor with an IC50 of 90 nM. It exhibits strong prophylactic effects in a pathogenic influenza model, inhibiting tumor growth and prolonging survival in a B16-OVA cells (s.c.) xenograft mice model. It is for research use only.
  • FPPS inhibitor with an IC50 of 90 nM
  • Exhibits strong prophylactic effects in pathogenic influenza model
  • Inhibits tumor growth and prolongs survival in xenograft mice model