Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TH-Z93 | 2260887-09-8 | C12H22N2O7P2 | 50 MG
SDP

Catalog No. 5000375066
Encompass_Preferred

TH-Z93 is a lipophilic bisphosphonate that functions as a potent FPPS inhibitor with an IC50 of 90 nM. This compound is suitable for various research applications requiring FPPS inhibition.

  • Potent FPPS inhibitor (IC50: 90 nM)
  • Lipophilic bisphosphonate
  • Molecular weight: 368.26
  • Appearance: White to off-white solid
  • Purity: 98.41%
  • Long-term powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in water: 7.81 mg/mL (21.21 mM)

Catalog No. 50-003-75066 Supplier Medchemexpress LLC Supplier No. HY15016750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TH-Z93 is a lipophilic bisphosphonate that functions as a potent FPPS inhibitor with an IC50 of 90 nM. This compound is suitable for various research applications requiring FPPS inhibition.

  • Potent FPPS inhibitor (IC50: 90 nM)
  • Lipophilic bisphosphonate
  • Molecular weight: 368.26
  • Appearance: White to off-white solid
  • Purity: 98.41%
  • Long-term powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in water: 7.81 mg/mL (21.21 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.