Learn More
Medchemexpress LLC HY-116961 5mg Medchemexpress, TH1020 CAS:1841460-82-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1169615MG
Encompass_Preferred
Medchemexpress, HY-116961 5mg TH1020 CAS:1841460-82-9 TH1020 is a potent and selective toll-like receptor 5 (TLR5)/flagellin complex antagonist with an IC50 of 0.85 μM. TH1020 inhbits flagellin-induced TLR5 signaling. TH1020 is inactive against TLR2, TLR3, TLR4, TLR7 and TLR8. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9819