Learn More
Medchemexpress LLC TH1834 dihydrochloride | 2108830-09-5 | ≥99.0% | 641.63 | C33H42Cl2N6O3 | 5MG
SDP
Catalog No. 5000238626
Encompass_Preferred
TH1834 dihydrochloride is the dihydrochloride salt of TH1834, a small-molecule inhibitor of the Tip60 (KAT5) histone acetyltransferase used in cellular and biochemical research to probe histone acetylation, apoptosis, and DNA damage responses. The compound has chemical formula C33H42Cl2N6O3, molecular weight 641.63, and CAS 2108830-09-5; it is supplied as a high-purity research reagent.
  • Specific Tip60 (KAT5) histone acetyltransferase inhibitor.
  • Dihydrochloride salt form for improved solubility.
  • High purity suitable for research (≥99.0%).
  • Supplied as a small research sample (5 mg).
  • Used in studies of apoptosis and DNA damage responses.
  • For research use only; not for clinical or diagnostic applications.
Catalog No. 50-002-38626 Supplier Medchemexpress LLC Supplier No. HY123604A5MG
TH1834 dihydrochloride is the dihydrochloride salt of TH1834, a small-molecule inhibitor of the Tip60 (KAT5) histone acetyltransferase used in cellular and biochemical research to probe histone acetylation, apoptosis, and DNA damage responses. The compound has chemical formula C33H42Cl2N6O3, molecular weight 641.63, and CAS 2108830-09-5; it is supplied as a high-purity research reagent.
  • Specific Tip60 (KAT5) histone acetyltransferase inhibitor.
  • Dihydrochloride salt form for improved solubility.
  • High purity suitable for research (≥99.0%).
  • Supplied as a small research sample (5 mg).
  • Used in studies of apoptosis and DNA damage responses.
  • For research use only; not for clinical or diagnostic applications.