Learn More
Aviva Systems Biology Corp TH1L Antibody - N-terminal region (ARP57805_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TH1L. It was validated on Western Blot. TH1L,TH1-like (Drosophila),TH1; NELF-C; NELF-D; HSPC130; TH1L,negative elongation factor C/D,negative elongation factor C/D,NELF-C/D,TH1-like protein,trihydrophobin 1,TH1 drosophila homolog,negative elongation factor proteins C and DThe immunogen is a synthetic peptide directed towards the N terminal region of human TH1L. Alternate names: HSPC130, NELF-C, NELF-D, TH1, TH1LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-387