Learn More
Selleck Chemical LLC TH287 10mg 1609960-30-6
SDP
Supplier:  Selleck Chemical LLC S763110MG
Encompass_Preferred
TH287 is a potent and selective MTH1 (NUDT1) inhibitor with IC50 of 0.8 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5044