Learn More
TARGETMOL CHEMICALS INC TH34 10MG
SDP
Catalog No. 502576692
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TH34 is an HDAC6/8/10 inhibitor (IC50s 4.6/1.9/7.7 uM). It shows high selectivity over HDAC1/2/3. purity: 98%
Catalog No. 50-257-6692 Supplier TARGETMOL CHEMICALS INC Supplier No. T533210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TH34 is an HDAC6/8/10 inhibitor (IC50s 4.6/1.9/7.7 uM). It shows high selectivity over HDAC1/2/3. purity: 98%