Learn More
Medchemexpress LLC HY-111818 10mg Medchemexpress, TH34 CAS:2196203-96-8 Purity:>98%
SDP
Catalog No. 501968973
Encompass_Preferred
Medchemexpress, HY-111818 10mg TH34 CAS:2196203-96-8 TH34, an HDAC6/8/10 inhibitor with IC50s of 4.6 μM, 1.9 μM, and 7.7 μM respectively, shows high selectivity over HDAC1/2/3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8973 Supplier Medchemexpress LLC Supplier No. HY11181810MG
Medchemexpress, HY-111818 10mg TH34 CAS:2196203-96-8 TH34, an HDAC6/8/10 inhibitor with IC50s of 4.6 μM, 1.9 μM, and 7.7 μM respectively, shows high selectivity over HDAC1/2/3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.