Learn More
Medchemexpress LLC HY-111818 5mg Medchemexpress, TH34 CAS:2196203-96-8 Purity:>98%
SDP
Catalog No. 501932399
Encompass_Preferred
Medchemexpress, HY-111818 5mg TH34 CAS:2196203-96-8 TH34, an HDAC6 / 8 / 10 inhibitor with IC 50 s of 4.6 μM, 1.9 μM, and 7.7 μM respectively, shows high selectivity over HDAC1/2/3 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2399 Supplier Medchemexpress LLC Supplier No. HY1118185MG
Medchemexpress, HY-111818 5mg TH34 CAS:2196203-96-8 TH34, an HDAC6 / 8 / 10 inhibitor with IC 50 s of 4.6 μM, 1.9 μM, and 7.7 μM respectively, shows high selectivity over HDAC1/2/3 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.