Learn More
Selleck Chemical LLC TH5487 5mg
SDP
Catalog No. 502173430
Encompass_Preferred
TH5487 is a selective active-site inhibitor of 8-oxoguanine DNA glycosylase 1 (OGG1) with IC50 of 342 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3430 Supplier Selleck Chemical LLC Supplier No. S89135MG
TH5487 is a selective active-site inhibitor of 8-oxoguanine DNA glycosylase 1 (OGG1) with IC50 of 342 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.