Learn More
Medchemexpress LLC HY-125276 5mg Medchemexpress, TH5487 CAS:2304947-71-3 Purity:>98%
SDP
Catalog No. 501969689
Encompass_Preferred
Medchemexpress, HY-125276 5mg TH5487 CAS:2304947-71-3 TH5487 is a potent 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor with an IC50 of 342 nM. TH5487 stops OGG1 from recognizing its DNA substrate, inhibits DNA repair and modifies OGG1 chromatin dynamics, which results in the inhibition of proinflammatory pathway genes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9689 Supplier Medchemexpress LLC Supplier No. HY1252765MG
Medchemexpress, HY-125276 5mg TH5487 CAS:2304947-71-3 TH5487 is a potent 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor with an IC50 of 342 nM. TH5487 stops OGG1 from recognizing its DNA substrate, inhibits DNA repair and modifies OGG1 chromatin dynamics, which results in the inhibition of proinflammatory pathway genes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.