Learn More
Eurofins Discoverx TH588, 5 mg. (92-1325)
Supplier:  Eurofins Discoverx 921325
Encompass
TH588 is a potent selective inhibitor of MTH1 (NUDT1). It can also induce DNA damage and triggers an ATM-p53-mediated death response and DNA repair in U2OS cells. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9837