Learn More
Medchemexpress LLC HY-12814 5mg Medchemexpress, TH588 CAS:1609960-31-7 Purity:>98%
SDP
Catalog No. 502081153
Encompass_Preferred
Medchemexpress, HY-12814 5mg TH588 CAS:1609960-31-7 TH588 is first-in-class nudix hydrolase family inhibitor that potently and selectively engage and inhibit the MTH1 (IC50= 5 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1153 Supplier Medchemexpress LLC Supplier No. HY128145MG
Medchemexpress, HY-12814 5mg TH588 CAS:1609960-31-7 TH588 is first-in-class nudix hydrolase family inhibitor that potently and selectively engage and inhibit the MTH1 (IC50= 5 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.