Learn More
AGRI PRO ENTERPRISES OF IOWA INC THA-MA 210 1CC SYRINGE
Catalog No. NC2824329
Encompass_Preferred
This 1 ML ThaMa 210 is a highly versatile automatic syringe designed for ultra precision and maximum ease-of-use. All parts of the syringe can be sterilized up to 125 °C/257 °F. Thermal-treated glass barrel. Adjustable from 0.1ml - 1.0ml.
Catalog No. NC2824329 Supplier AGRI PRO ENTERPRISES OF IOWA INC Supplier No. 173400
This 1 ML ThaMa 210 is a highly versatile automatic syringe designed for ultra precision and maximum ease-of-use. All parts of the syringe can be sterilized up to 125 °C/257 °F. Thermal-treated glass barrel. Adjustable from 0.1ml - 1.0ml.