Learn More
AdipoGen Thailandolide B (1 mg)
SDP
Supplier:  AdipoGen AGCN20160M001
Encompass_Preferred
Chemical. CAS: 944726-62-9. Formula: C27H34O8. MW: 484.6. Isolated from Penicillium sp. Antibiotic. Stereoisomer of chrodrimanin B, which is an insecticidal compound.
Catalog No. 50-205-3548