Learn More
Medchemexpress LLC Thailanstatin A cyclohexane diamine (formic) | 93.7% | 677.83 | 1 MG
SDP
Catalog No. 5000392262
Encompass_Preferred
Thailanstatin A cyclohexane diamine formic is an analog of Spliceostatin. It functions as a drug-linker conjugate for Antibody-Drug Conjugate (ADC) molecules. This product is for research use only and not intended for patient use.
  • Inhibits proliferation of N87 cells (IC50 0.33 μM)
  • Inhibits proliferation of BT474 cells (IC50 0.34 μM)
  • Inhibits proliferation of MDA-MB-361-DYT2 cells (IC50 0.88 μM)
  • Inhibits proliferation of MDA-MB-468 cells (IC50 0.27 μM)
  • Analog of Spliceostatin
Catalog No. 50-003-92262 Supplier Medchemexpress LLC Supplier No. HY160899A1MG
Add to Cart
Add to Cart
Thailanstatin A cyclohexane diamine formic is an analog of Spliceostatin. It functions as a drug-linker conjugate for Antibody-Drug Conjugate (ADC) molecules. This product is for research use only and not intended for patient use.
  • Inhibits proliferation of N87 cells (IC50 0.33 μM)
  • Inhibits proliferation of BT474 cells (IC50 0.34 μM)
  • Inhibits proliferation of MDA-MB-361-DYT2 cells (IC50 0.88 μM)
  • Inhibits proliferation of MDA-MB-468 cells (IC50 0.27 μM)
  • Analog of Spliceostatin