Learn More
Medchemexpress LLC HY-123937 5mg Medchemexpress, THAL-SNS-032 CAS:2139287-33-3 Purity:>98%
SDP
Catalog No. 501932586
Encompass_Preferred
Medchemexpress, HY-123937 5mg THAL-SNS-032 CAS:2139287-33-3 THAL-SNS-032 is a selective CDK9 degrader PROTAC consisting of a CDK-binding SNS-032 ligand linked to a thalidomide derivative that binds the E3 ubiquitin ligase Cereblon (CRBN) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2586 Supplier Medchemexpress LLC Supplier No. HY1239375MG
Medchemexpress, HY-123937 5mg THAL-SNS-032 CAS:2139287-33-3 THAL-SNS-032 is a selective CDK9 degrader PROTAC consisting of a CDK-binding SNS-032 ligand linked to a thalidomide derivative that binds the E3 ubiquitin ligase Cereblon (CRBN) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.