Learn More
Sigma Aldrich Fine Chemicals Biosciences ThalesNano CatCart holder with SS connection tube, L × diam. 30 mm × 4 mm
Catalog No. NC1629143
Encompass
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Catalog No. NC1629143 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. THS012191EA
CatCarts are suitable for use only with Thalesnano H-Cube reactors.