Learn More
Medchemexpress LLC Thalidomide-1-me,4-oh | 2244568-06-5 | ≥98.0% | 288.26 g·mol-1 | C14H12N2O5 | 100 MG
SDP
Catalog No. 5000280980
Encompass_Preferred
Thalidomide-1-Me,4-OH is a thalidomide analogue bearing a 4-hydroxy substituent and an N-1 methyl group. It serves as a cereblon/E3 ligase ligand scaffold for research into ubiquitin-mediated proteolysis and is supplied as a solid research reagent.
  • Small-molecule cereblon/E3 ligase ligand used in biochemical and cell-based assays
  • High purity suitable for research applications
  • Powder form with defined storage recommendations for long-term stability
  • Includes chemical identifiers for traceability, including CAS and molecular formula
  • Applicable for studies of targeted protein degradation and ligand design
Catalog No. 50-002-80980 Supplier Medchemexpress LLC Supplier No. HYW441376100MG
Thalidomide-1-Me,4-OH is a thalidomide analogue bearing a 4-hydroxy substituent and an N-1 methyl group. It serves as a cereblon/E3 ligase ligand scaffold for research into ubiquitin-mediated proteolysis and is supplied as a solid research reagent.
  • Small-molecule cereblon/E3 ligase ligand used in biochemical and cell-based assays
  • High purity suitable for research applications
  • Powder form with defined storage recommendations for long-term stability
  • Includes chemical identifiers for traceability, including CAS and molecular formula
  • Applicable for studies of targeted protein degradation and ligand design