Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-1-me-5-nh2 | 1468761-60-5 | >98.0% | 287.27 g·mol-1 | C14H13N3O4 | 50 MG
SDP

Catalog No. 5000284624
Encompass_Preferred

Thalidomide-1-Me-5-NH2 is an aminated thalidomide analogue used as a cereblon E3 ligase ligand and building block for PROTAC design and related chemical biology research. It is intended for derivatization at the amine for linker attachment and is supplied as a high-purity research reagent suitable for synthetic applications.

  • Aminated thalidomide analogue enabling linker derivatization
  • Serves as a cereblon E3 ligase ligand for PROTAC design
  • High purity for research applications (>98.0%)
  • Supplied in small milligram-scale quantities suitable for synthesis (50 mg)
  • Molecular formula C14H13N3O4, molecular weight 287.27 g·mol-1
  • CAS number 1468761-60-5 for unambiguous identification

Catalog No. 50-002-84624 Supplier Medchemexpress LLC Supplier No. HYW58682150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-1-Me-5-NH2 is an aminated thalidomide analogue used as a cereblon E3 ligase ligand and building block for PROTAC design and related chemical biology research. It is intended for derivatization at the amine for linker attachment and is supplied as a high-purity research reagent suitable for synthetic applications.

  • Aminated thalidomide analogue enabling linker derivatization
  • Serves as a cereblon E3 ligase ligand for PROTAC design
  • High purity for research applications (>98.0%)
  • Supplied in small milligram-scale quantities suitable for synthesis (50 mg)
  • Molecular formula C14H13N3O4, molecular weight 287.27 g·mol-1
  • CAS number 1468761-60-5 for unambiguous identification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.